Anti-EMC3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB122786-100
ABCAAB122786-100EA 857 EUR
ABCAAB122786-100
Anti-EMC3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EMC3 (ER membrane protein complex subunit 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR