Anti-NUP107 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB122883-200EA 797 EUR
ABCAAB122883-200
Anti-NUP107 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NUP107 (nucleoporin 107kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR