Anti-FGF23 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB123502-200EA 840 EUR
ABCAAB123502-200
Anti-FGF23 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FGF23 (fibroblast growth factor 23)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR