Anti-THY1 Mouse Monoclonal Antibody [clone: 5E10]

Lieferant: Abcam
ABCAAB123511-100EA 852 EUR
ABCAAB123511-100
Anti-THY1 Mouse Monoclonal Antibody [clone: 5E10]
Antikörper
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: 50000000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR