Anti-VIPR1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB123517-200EA 539 EUR
ABCAAB123517-200
Anti-VIPR1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VIPR1 (vasoactive intestinal peptide receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR