Anti-PTPRC Mouse monoclonal antibody [clone: HI30]
Lieferant: Abcam
ABCAAB123522-100EA 771 EUR
ABCAAB123522-100
Anti-PTPRC Mouse monoclonal antibody [clone: HI30]
Type: Primary
Antigen: PTPRC (protein tyrosine phosphatase, receptor type, C)
Clonality: monoclonal
Clone: HI30
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen