Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody

Lieferant: Abcam
AB123946-100
ABCAAB123946-100EA 896 EUR
ABCAAB123946-100
Anti-Macrophage Scavenger Receptor I Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Macrophage Scavenger Receptor I
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR