Anti-HSPA13 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB124208-100EA 816 EUR
ABCAAB124208-100
Anti-HSPA13 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSPA13 (heat shock protein 70kDa family, member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR