Anti-CDK8 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB124218-100EA 840 EUR
ABCAAB124218-100
Anti-CDK8 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: CDK8 (cyclin-dependent kinase 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR