Anti-MARK4 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB124267-50EA 708 EUR
ABCAAB124267-50
Anti-MARK4 Rabbit polyclonal antibody
Type: Primary
Antigen: MARK4 (MAP/microtubule affinity-regulating kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR