Anti-STIL Rabbit Polyclonal Antibody

Lieferant: Abcam
AB124322-100
ABCAAB124322-100EA 836 EUR
ABCAAB124322-100
Anti-STIL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STIL (SCL/TAL1 interrupting locus)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR