Anti-PAX8 Mouse Monoclonal Antibody [clone: BC12]

Lieferant: Abcam
ABCAAB124445-100EA 840 EUR
ABCAAB124445-100
Anti-PAX8 Mouse Monoclonal Antibody [clone: BC12]
Antikörper
Type: Primary
Antigen: PAX8 (paired box 8)
Clonality: monoclonal
Clone: BC12
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR