Anti-Hepatitis C Virus NS4A Mouse Monoclonal Antibody (Biotin) [clone: S4-13]

Lieferant: Abcam
ABCAAB124526-50EA 815 EUR
ABCAAB124526-50
Anti-Hepatitis C Virus NS4A Mouse Monoclonal Antibody (Biotin) [clone: S4-13]
Antikörper
Type: Primary
Antigen: Hepatitis C Virus NS4A
Clonality: monoclonal
Clone: S4-13
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR