Anti-ERAP1 Rabbit Monoclonal Antibody [clone: EPR6069]

Lieferant: Abcam
AB124669-100
ABCAAB124669-100EA 825 EUR
ABCAAB124669-100
Anti-ERAP1 Rabbit Monoclonal Antibody [clone: EPR6069]
Antikörper
Type: Primary
Antigen: ERAP1 (endoplasmic reticulum aminopeptidase 1)
Clonality: monoclonal
Clone: EPR6069
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR