Anti-DP1 Rabbit Monoclonal Antibody [clone: EPR4959]

Lieferant: Abcam
AB124678-100
ABCAAB124678-100EA 827 EUR
ABCAAB124678-100
Anti-DP1 Rabbit Monoclonal Antibody [clone: EPR4959]
Antikörper
Type: Primary
Antigen: DP1
Clonality: monoclonal
Clone: EPR4959
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR