Anti-ARL4A Rabbit Monoclonal Antibody [clone: EPR6086]

Lieferant: Abcam
ABCAAB124690-100EA 778 EUR
ABCAAB124690-100
Anti-ARL4A Rabbit Monoclonal Antibody [clone: EPR6086]
Antikörper
Type: Primary
Antigen: ARL4A (ADP-ribosylation factor-like 4A)
Clonality: monoclonal
Clone: EPR6086
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR