Anti-DUX4 Rabbit Monoclonal Antibody [clone: E55]

Lieferant: Abcam
ABCAAB124699-100EA 864 EUR
ABCAAB124699-100
Anti-DUX4 Rabbit Monoclonal Antibody [clone: E55]
Antikörper
Type: Primary
Antigen: DUX4 (double homeobox 4)
Clonality: monoclonal
Clone: E55
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR