Anti-REM1 Rabbit Monoclonal Antibody [clone: EP4482]

Lieferant: Abcam
ABCAAB124703-100EA 765 EUR
ABCAAB124703-100
Anti-REM1 Rabbit Monoclonal Antibody [clone: EP4482]
Antikörper
Type: Primary
Antigen: REM1 (RAS (RAD and GEM)-like GTP-binding 1)
Clonality: monoclonal
Clone: EP4482
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR