Anti-DBF4 Rabbit monoclonal antibody unconjugated [clone: EPR5635]

Lieferant: Abcam
ABCAAB124707-100EA 779 EUR
ABCAAB124707-100
Anti-DBF4 Rabbit monoclonal antibody unconjugated [clone: EPR5635]
Antikörper
Type: Primary
Antigen: DBF4 (DBF4 zinc finger)
Clonality: monoclonal
Clone: EPR5635
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR