Anti-COPS7B Rabbit Monoclonal Antibody [clone: EPR6465]

Lieferant: Abcam
AB124718-100
ABCAAB124718-100EA 980 EUR
ABCAAB124718-100
Anti-COPS7B Rabbit Monoclonal Antibody [clone: EPR6465]
Antikörper
Type: Primary
Antigen: COPS7B (COP9 signalosome subunit 7B)
Clonality: monoclonal
Clone: EPR6465
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR