Anti-PLCB3 Rabbit Monoclonal Antibody [clone: EPR5951]
Lieferant: Abcam
ABCAAB124735-100EA 854 EUR
ABCAAB124735-100
Anti-PLCB3 Rabbit Monoclonal Antibody [clone: EPR5951]
Antikörper
Type: Primary
Antigen: PLCB3 (phospholipase C, beta 3 (phosphatidylinositol-specific))
Clonality: monoclonal
Clone: EPR5951
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen