Anti-SUN1 Rabbit Monoclonal Antibody [clone: EPR6554]

Lieferant: Abcam
ABCAAB124770-100EA 886 EUR
ABCAAB124770-100
Anti-SUN1 Rabbit Monoclonal Antibody [clone: EPR6554]
Antikörper
Type: Primary
Antigen: SUN1 (Sad1 and UNC84 domain containing 1)
Clonality: monoclonal
Clone: EPR6554
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR