Anti-S100A4 Rabbit Monoclonal Antibody [clone: EPR2761]

Lieferant: Abcam
AB124805-100
ABCAAB124805-100EA 997 EUR
ABCAAB124805-100
Anti-S100A4 Rabbit Monoclonal Antibody [clone: EPR2761]
Antikörper
Type: Primary
Antigen: S100A4 (S100 calcium binding protein A4)
Clonality: monoclonal
Clone: EPR2761
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR