Anti-SERPINC1 Rabbit Monoclonal Antibody [clone: EPR5371]

Lieferant: Abcam
ABCAAB124808-100EA 873 EUR
ABCAAB124808-100
Anti-SERPINC1 Rabbit Monoclonal Antibody [clone: EPR5371]
Antikörper
Type: Primary
Antigen: SERPINC1 (serpin peptidase inhibitor, clade C (antithrombin), member 1)
Clonality: monoclonal
Clone: EPR5371
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR