Anti-FADD Rabbit Monoclonal Antibody [clone: EPR5030]

Lieferant: Abcam
AB124812-100
ABCAAB124812-100EA 908 EUR
ABCAAB124812-100
Anti-FADD Rabbit Monoclonal Antibody [clone: EPR5030]
Antikörper
Type: Primary
Antigen: FADD (Fas (TNFRSF6)-associated via death domain)
Clonality: monoclonal
Clone: EPR5030
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR