Anti-CDK6 Rabbit monoclonal antibody unconjugated [clone: EPR4515]

Lieferant: Abcam
ABCAAB124821-100EA 944 EUR
ABCAAB124821-100
Anti-CDK6 Rabbit monoclonal antibody unconjugated [clone: EPR4515]
Antikörper
Type: Primary
Antigen: CDK6 (cyclin-dependent kinase 6)
Clonality: monoclonal
Clone: EPR4515
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR