Anti-CD209 Rabbit Monoclonal Antibody [clone: EPR5588]

Lieferant: Abcam
AB124828-100
ABCAAB124828-100EA 902 EUR
ABCAAB124828-100
Anti-CD209 Rabbit Monoclonal Antibody [clone: EPR5588]
Antikörper
Type: Primary
Antigen: CD209 (CD209 molecule)
Clonality: monoclonal
Clone: EPR5588
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR