Anti-GPC3 Rabbit Monoclonal Antibody [clone: EPR5547]

Lieferant: Abcam
AB124829-100
ABCAAB124829-100EA 876 EUR
ABCAAB124829-100
Anti-GPC3 Rabbit Monoclonal Antibody [clone: EPR5547]
Antikörper
Type: Primary
Antigen: GPC3 (glypican 3)
Clonality: monoclonal
Clone: EPR5547
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR