Anti-TMOD2 Rabbit Monoclonal Antibody [clone: EPR6353]

Lieferant: Abcam
ABCAAB124833-100EA 818 EUR
ABCAAB124833-100
Anti-TMOD2 Rabbit Monoclonal Antibody [clone: EPR6353]
Antikörper
Type: Primary
Antigen: TMOD2 (tropomodulin 2 (neuronal))
Clonality: monoclonal
Clone: EPR6353
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR