Anti-TYROBP Rabbit Monoclonal Antibody [clone: EPR5173]

Lieferant: Abcam
ABCAAB124834-100EA 857 EUR
ABCAAB124834-100
Anti-TYROBP Rabbit Monoclonal Antibody [clone: EPR5173]
Antikörper
Type: Primary
Antigen: TYROBP (TYRO protein tyrosine kinase binding protein)
Clonality: monoclonal
Clone: EPR5173
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR