Anti-CPN1 Rabbit Monoclonal Antibody [clone: EPR6229]

Lieferant: Abcam
ABCAAB124874-100EA 761 EUR
ABCAAB124874-100
Anti-CPN1 Rabbit Monoclonal Antibody [clone: EPR6229]
Antikörper
Type: Primary
Antigen: CPN1 (carboxypeptidase N, polypeptide 1)
Clonality: monoclonal
Clone: EPR6229
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR