Anti-MEOX2 Rabbit Monoclonal Antibody [clone: EPR5568]

Lieferant: Abcam
AB124876-100
ABCAAB124876-100EA 816 EUR
ABCAAB124876-100
Anti-MEOX2 Rabbit Monoclonal Antibody [clone: EPR5568]
Antikörper
Type: Primary
Antigen: MEOX2 (mesenchyme homeobox 2)
Clonality: monoclonal
Clone: EPR5568
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR