Anti-SPTBN1 Rabbit Monoclonal Antibody [clone: EPR5869]

Lieferant: Abcam
ABCAAB124888-100EA 853 EUR
ABCAAB124888-100
Anti-SPTBN1 Rabbit Monoclonal Antibody [clone: EPR5869]
Antikörper
Type: Primary
Antigen: SPTBN1 (spectrin, beta, non-erythrocytic 1)
Clonality: monoclonal
Clone: EPR5869
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR