Anti-Mitochondrial Ferritin Rabbit Monoclonal Antibody [clone: EPR1797]

Lieferant: Abcam
ABCAAB124889-100EA 778 EUR
ABCAAB124889-100
Anti-Mitochondrial Ferritin Rabbit Monoclonal Antibody [clone: EPR1797]
Antikörper
Type: Primary
Antigen: Mitochondrial Ferritin
Clonality: monoclonal
Clone: EPR1797
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR