Anti-NDEL1 Rabbit Monoclonal Antibody [clone: EPR5068]

Lieferant: Abcam
ABCAAB124895-100EA 853 EUR
ABCAAB124895-100
Anti-NDEL1 Rabbit Monoclonal Antibody [clone: EPR5068]
Antikörper
Type: Primary
Antigen: NDEL1 (nudE neurodevelopment protein 1-like 1)
Clonality: monoclonal
Clone: EPR5068
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR