Anti-Munc18 Rabbit Monoclonal Antibody [clone: EPR4850]

Lieferant: Abcam
ABCAAB124920-100EA 838 EUR
ABCAAB124920-100
Anti-Munc18 Rabbit Monoclonal Antibody [clone: EPR4850]
Antikörper
Type: Primary
Antigen: Munc18
Clonality: monoclonal
Clone: EPR4850
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR