Anti-Lipoamide Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR6634]

Lieferant: Abcam
ABCAAB124926-100EA 778 EUR
ABCAAB124926-100
Anti-Lipoamide Dehydrogenase Rabbit Monoclonal Antibody [clone: EPR6634]
Antikörper
Type: Primary
Antigen: Lipoamide Dehydrogenase
Clonality: monoclonal
Clone: EPR6634
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR