Anti-ICOSLG Rabbit Monoclonal Antibody [clone: EPR6071]

Lieferant: Abcam
AB124972-100
ABCAAB124972-100EA 892 EUR
ABCAAB124972-100
Anti-ICOSLG Rabbit Monoclonal Antibody [clone: EPR6071]
Antikörper
Type: Primary
Antigen: ICOSLG (inducible T-cell co-stimulator ligand)
Clonality: monoclonal
Clone: EPR6071
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR