Anti-FYN Rabbit Monoclonal Antibody [clone: EPR5500]

Lieferant: Abcam
AB125016-100
ABCAAB125016-100EA 915 EUR
ABCAAB125016-100
Anti-FYN Rabbit Monoclonal Antibody [clone: EPR5500]
Antikörper
Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: monoclonal
Clone: EPR5500
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR