Anti-MRC1 Rabbit Monoclonal Antibody [clone: EPR6828]
Lieferant: Abcam
ABCAAB125028-100EA 874 EUR
ABCAAB125028-100
Anti-MRC1 Rabbit Monoclonal Antibody [clone: EPR6828]
Antikörper
Type: Primary
Antigen: MRC1 (mannose receptor, C type 1)
Clonality: monoclonal
Clone: EPR6828
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen