Anti-MC1R Rabbit Monoclonal Antibody [clone: EPR6530]

Lieferant: Abcam
AB125031-100
ABCAAB125031-100EA 908 EUR
ABCAAB125031-100
Anti-MC1R Rabbit Monoclonal Antibody [clone: EPR6530]
Antikörper
Type: Primary
Antigen: MC1R (melanocortin 1 receptor (alpha melanocyte stimulating hormone receptor))
Clonality: monoclonal
Clone: EPR6530
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR