Anti-AQP3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
AB125045-100
ABCAAB125045-100EA 902 EUR
ABCAAB125045-100
Anti-AQP3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: AQP3 (aquaporin 3 (Gill blood group))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR