Anti-CD68 Rabbit polyclonal antibody

Lieferant: Abcam
ABCAAB125047-1EA 1300 EUR
ABCAAB125047-1
Anti-CD68 Rabbit polyclonal antibody
Type: Primary
Antigen: CD68 (CD68 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR