Anti-GPX4 Rabbit Monoclonal Antibody [clone: EPNCIR144]

Lieferant: Abcam
AB125066-100
ABCAAB125066-100EA 1040 EUR
ABCAAB125066-100
Anti-GPX4 Rabbit Monoclonal Antibody [clone: EPNCIR144]
Antikörper
Type: Primary
Antigen: GPX4 (glutathione peroxidase 4)
Clonality: monoclonal
Clone: EPNCIR144
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR