Anti-RIPK1 Rabbit Monoclonal Antibody [clone: EPR4689]
Lieferant: Abcam
ABCAAB125072-100EA 906 EUR
ABCAAB125072-100
Anti-RIPK1 Rabbit Monoclonal Antibody [clone: EPR4689]
Antikörper
Type: Primary
Antigen: RIPK1 (receptor (TNFRSF)-interacting serine-threonine kinase 1)
Clonality: monoclonal
Clone: EPR4689
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen