Anti-MYF5 Rabbit Monoclonal Antibody [clone: EPR4899]

Lieferant: Abcam
AB125078-100
ABCAAB125078-100EA 918 EUR
ABCAAB125078-100
Anti-MYF5 Rabbit Monoclonal Antibody [clone: EPR4899]
Antikörper
Type: Primary
Antigen: MYF5 (myogenic factor 5)
Clonality: monoclonal
Clone: EPR4899
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR