Anti-SCRIBBLE Rabbit Monoclonal Antibody [clone: EPR4140]

Lieferant: Abcam
ABCAAB125080-100EA 835 EUR
ABCAAB125080-100
Anti-SCRIBBLE Rabbit Monoclonal Antibody [clone: EPR4140]
Antikörper
Type: Primary
Antigen: SCRIBBLE
Clonality: monoclonal
Clone: EPR4140
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR