Anti-FYTTD1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB125082-100EA 797 EUR
ABCAAB125082-100
Anti-FYTTD1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FYTTD1 (forty-two-three domain containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR