Anti-USF2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB125184-100EA 835 EUR
ABCAAB125184-100
Anti-USF2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: USF2 (upstream transcription factor 2, c-fos interacting)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR