Anti-TRIM27 Mouse Monoclonal Antibody [clone: RF5R]

Lieferant: Abcam
ABCAAB125244-100EA 852 EUR
ABCAAB125244-100
Anti-TRIM27 Mouse Monoclonal Antibody [clone: RF5R]
Antikörper
Type: Primary
Antigen: TRIM27 (tripartite motif containing 27)
Clonality: monoclonal
Clone: RF5R
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR